5-Chlor-2-thenoesäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Chlor-2-thiophencarbonsäure
, 5-Chlorthiophen-2-carbonsäure
A15209.22EA
341
EUR
A15209.22
5-Chlor-2-thenoesäure ≥98%
5-Chlor-2-thenoesäure
Formel:
C₅H₃ClO₂S Molecular Weight: 162,6 g/mol Schmelzpunkt: 149…152 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041426 CAS-Nummer: 24065-33-6 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...