L(-)-Prolin
Synonyme:
(S)-(-)-Pyrrolidin-2-carbonsäure
, H-Pro-OH
, (2S)-(-)-Pyrrolidin-2-carbonsäure
, (S)-(-)-1,3,4,5-Tetrahydro-2H-pyrrol- 2-carbonsäure
, (S)-(-)-Prolin
, (S)-(-)-Azolidin-2-carbonsäure
EDQMP3350000EA
235
EUR
EDQMP3350000
L(-)-Prolin
L(-)-Prolin
Formel:
C₅H₉NO₂ Molecular Weight: 115,13 g/mol Siedepunkt: 245 °C (1013 hPa) Schmelzpunkt: 220…222 °C Dichte: 1,35 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064318 CAS-Nummer: 147-85-3 EINECS: 205-702-2 Merck Index: 12,07963 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...