VWR®, ASTM D 6584 standard set of 7 CRMs
Lieferant: Avantor
VWR®
91625.001EA 793 EUR
91625.001
VWR®, ASTM D 6584 standard set of 7 CRMs
Standards Organische Standards
Glycerin, Monoolein, 1,3-Diolein, Triolein, (S)-(-)-1,2,4-Butanetriol, Tricaprin, MSTFA.
Bitte beachten Highly Flammable.