3-Hydroxyisonicotinsäure 97%
|
|
BLDPBD9932-5GEA
37
EUR
BLDPBD9932-5G
BLDPBD9932-25G
BLDPBD9932-1G
BLDPBD9932-10G
BLDPBD9932-100G
3-Hydroxyisonicotinsäure 97%
3-Hydroxyisonicotinsäure
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel:
C₆H₅NO₃ Molecular Weight: 139,11 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234165 CAS-Nummer: 10128-71-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...