4-Methylbenzoesäureanhydrid 95%

BLDPBD98981-1GEA 25 EUR
BLDPBD98981-1G BLDPBD98981-25G BLDPBD98981-5G
4-Methylbenzoesäureanhydrid 95%
4-Methylbenzoic anhydride

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₆H₁₄O₃
Molecular Weight: 254,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00048100
CAS-Nummer: 13222-85-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR