4-Methyl-2-(3-pyridinyl)-5-thiazolcarbonsäure 98%

BLDPBD98734-250MGEA 18 EUR
BLDPBD98734-250MG
4-Methyl-2-(3-pyridinyl)-5-thiazolcarbonsäure 98%
4-Methyl-2-(3-pyridinyl)-5-thiazolcarbonsäure

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formel: C₁₀H₈N₂O₂S
Molecular Weight: 220,25 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00171753
CAS-Nummer: 39091-01-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR