5-Fluornicotinsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD9853-25GEA 56 EUR
BLDPBD9853-25G BLDPBD9853-5G BLDPBD9853-10G BLDPBD9853-1G
5-Fluornicotinsäure 98%
5-Fluornicotinsäure
Formel: C₆H₄FNO₂
Molecular Weight: 141,10 g/mol
MDL: MFCD01318537
CAS-Nummer: 402-66-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR