4-Methoxybenzolsulfonamid 98%
|
|
BLDPBD98486-25GEA
112
EUR
BLDPBD98486-25G
BLDPBD98486-100G
BLDPBD98486-5G
BLDPBD98486-1G
4-Methoxybenzolsulfonamid 98%
4-Methoxybenzolsulfonamid
Storage condition: Sealed in dry, Room Temperature
Formel:
C₇H₉NO₃S Molecular Weight: 187,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00025392 CAS-Nummer: 1129-26-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...