4-Iodo-2-methylpyridine 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD98075-100MGEA 34 EUR
BLDPBD98075-100MG BLDPBD98075-250MG
4-Iodo-2-methylpyridine 98%
4-Iodo-2-methylpyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₆H₆IN
Molecular Weight: 219,023 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09702484
CAS-Nummer: 22282-65-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR