4-Ethoxybenzylamin 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD96900-5GEA 42 EUR
BLDPBD96900-5G BLDPBD96900-1G BLDPBD96900-100G BLDPBD96900-10G BLDPBD96900-25G
4-Ethoxybenzylamin 95%
4-Ethoxybenzylamin

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₉H₁₃NO
Molecular Weight: 151,2056 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD02225920
CAS-Nummer: 6850-60-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR