4-(Chloromethyl)benzaldehyde 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD96358-1GEA 23 EUR
BLDPBD96358-0.025G BLDPBD96358-1G BLDPBD96358-5G BLDPBD96358-250MG BLDPBD96358-10G BLDPBD96358-25G
4-(Chloromethyl)benzaldehyde 95%
4-(Chloromethyl)benzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₇ClO
Molecular Weight: 154,594 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00191396
CAS-Nummer: 73291-09-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR