4'-Chlor-2-cyanacetanilid 98+%

Lieferant: BLD PHARMATECH GMBH

BLDPBD95701-5GEA 228 EUR
BLDPBD95701-5G BLDPBD95701-1G BLDPBD95701-250MG BLDPBD95701-100MG
4'-Chlor-2-cyanacetanilid 98+%
4'-Chlor-2-cyanacetanilid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₉H₇ClN₂O
Molecular Weight: 194,6177 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00036015
CAS-Nummer: 17722-17-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR