4-Brom-5-chlor-2-nitroanilin 95%

BLDPBD95114-100GEA 700 EUR
BLDPBD95114-100G BLDPBD95114-5G BLDPBD95114-10G BLDPBD95114-250MG BLDPBD95114-25G BLDPBD95114-1G
4-Brom-5-chlor-2-nitroanilin 95%
4-Brom-5-chlor-2-nitroanilin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₆H₄BrClN₂O₂
Molecular Weight: 251,4652 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11100159
CAS-Nummer: 827-33-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR