4,4-Dimethylcyclohexanol 97%

BLDPBD92235-5GEA 125 EUR
BLDPBD92235-5G BLDPBD92235-25G BLDPBD92235-1G BLDPBD92235-250MG
4,4-Dimethylcyclohexanol 97%
4,4-Dimethylcyclohexanol

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₁₆O
Molecular Weight: 128,2100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00101954
CAS-Nummer: 932-01-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR