4-(1,3-Oxazol-5-yl)anilin 97%

BLDPBD89441-250MGEA 17 EUR
BLDPBD89441-250MG BLDPBD89441-1G BLDPBD89441-5G
4-(1,3-Oxazol-5-yl)anilin 97%
4-(1,3-Oxazol-5-yl)anilin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₉H₈N₂O
Molecular Weight: 160,1700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00052192
CAS-Nummer: 1008-95-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR