3-Cyanpyridin-N-oxid 95+%

Lieferant: BLD PHARMATECH GMBH

BLDPBD89090-100MGEA 34 EUR
BLDPBD89090-100MG BLDPBD89090-250MG BLDPBD89090-1G BLDPBD89090-5G
3-Cyanpyridin-N-oxid 95+%
3-Cyanpyridin-N-oxid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₄N₂O
Molecular Weight: 120,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00129037
CAS-Nummer: 149060-64-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR