3-Methoxydiphenylamin 97%
|
|
BLDPBD87794-5GEA
9
EUR
BLDPBD87794-5G
BLDPBD87794-100G
BLDPBD87794-500G
BLDPBD87794-25G
BLDPBD87794-10G
3-Methoxydiphenylamin 97%
3-Methoxydiphenylamin
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₁₃H₁₃NO Molecular Weight: 199,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008383 CAS-Nummer: 101-16-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...