3-Methoxyzimtsäure 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD87792-5GEA 11 EUR
BLDPBD87792-5G BLDPBD87792-100G BLDPBD87792-25G
3-Methoxyzimtsäure 97%
3-Methoxyzimtsäure

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₀H₁₀O₃
Molecular Weight: 178,19 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00004386
CAS-Nummer: 6099-04-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR