Monomethyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate 98%
|
|
BLDPBD8754-10GEA
33
EUR
BLDPBD8754-10G
BLDPBD8754-500G
BLDPBD8754-100G
BLDPBD8754-25G
BLDPBD8754-5G
Monomethyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate 98%
Monomethyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate
Storage condition: Sealed in dry, Room Temperature
Formel:
C₁₆H₁₆N₂O₆ Molecular Weight: 332,31 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00475478 CAS-Nummer: 74936-72-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...