3-(Dimethylamino)phenol 97%

Synonyme: N,N-Dimethyl-3-hydroxyanilin

BLDPBD86493-100GEA 101 EUR
BLDPBD86493-100G BLDPBD86493-10G BLDPBD86493-5G BLDPBD86493-500G BLDPBD86493-25G
3-(Dimethylamino)phenol 97%
3-(Dimethylamino)phenol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₁₁NO
Molecular Weight: 137,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002264
CAS-Nummer: 99-07-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR