3-(Hydroxymethyl)benzonitril 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD86328-5GEA 46 EUR
BLDPBD86328-5G BLDPBD86328-10G BLDPBD86328-1G BLDPBD86328-25G
3-(Hydroxymethyl)benzonitril 98%
3-(Hydroxymethyl)benzonitril

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₇NO
Molecular Weight: 133,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01321087
CAS-Nummer: 874-97-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR