L-Glutaminsäure-5-methylester 97%
|
|
BLDPBD8630-25GEA
11
EUR
BLDPBD8630-25G
BLDPBD8630-100G
BLDPBD8630-1G
BLDPBD8630-10G
BLDPBD8630-1000G
BLDPBD8630-500G
BLDPBD8630-5G
L-Glutaminsäure-5-methylester 97%
L-Glutaminsäure-5-methylester
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₆H₁₁NO₄ Molecular Weight: 161,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002632 CAS-Nummer: 1499-55-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...