3-Chlor-N,N-dimethylanilin 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD86136-5GEA 57 EUR
BLDPBD86136-5G BLDPBD86136-25G BLDPBD86136-1G
3-Chlor-N,N-dimethylanilin 95%
3-Chlor-N,N-dimethylanilin

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₁₀ClN
Molecular Weight: 155,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00018456
CAS-Nummer: 6848-13-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR