Fmoc-L-phenylalanin 98%

Synonyme: N-α-Fmoc-L-PhenylalanineN-(9-Fluorenylmethoxycarbonyl)-L-phenylalanineFmoc-Phe-OH

BLDPBD8605-25GEA 13 EUR
BLDPBD8605-25G BLDPBD8605-10G BLDPBD8605-1000G BLDPBD8605-500G BLDPBD8605-5G BLDPBD8605-100G
Fmoc-L-phenylalanin 98%
Fmoc-L-phenylalanin

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₂₄H₂₁NO₄
Molecular Weight: 387,4 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00037128
CAS-Nummer: 35661-40-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR