3-Chlor-5-fluorphenylboronsäure 97%
|
|
BLDPBD85925-25GEA
167
EUR
BLDPBD85925-25G
BLDPBD85925-250MG
BLDPBD85925-5G
BLDPBD85925-10G
BLDPBD85925-1G
3-Chlor-5-fluorphenylboronsäure 97%
3-Chlor-5-fluorphenylboronsäure
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₆H₅BClFO₂ Molecular Weight: 174,37 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06801741 CAS-Nummer: 328956-61-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...