3-Chlor-4-hydroxyphenylboronsäurepinakolester 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD85822-5GEA 547 EUR
BLDPBD85822-5G BLDPBD85822-1G BLDPBD85822-250MG
3-Chlor-4-hydroxyphenylboronsäurepinakolester 98%
3-Chlor-4-hydroxyphenylboronsäurepinakolester

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₂H₁₆BClO₃
Molecular Weight: 254,5200 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09260490
CAS-Nummer: 629658-06-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR