N(α)-Boc-L-histidin 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD8521-100GEA 144 EUR
BLDPBD8521-100G BLDPBD8521-10G BLDPBD8521-25G BLDPBD8521-500G BLDPBD8521-5G
N(α)-Boc-L-histidin 98%
N(α)-Boc-L-histidin

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₁H₁₇N₃O₄
Molecular Weight: 255,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00065576
CAS-Nummer: 17791-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR