Diethyl-aminomalonat Hydrochlorid 98%
|
|
BLDPBD8498-500GEA
130
EUR
BLDPBD8498-500G
BLDPBD8498-1G
BLDPBD8498-25G
BLDPBD8498-100G
BLDPBD8498-5G
BLDPBD8498-1000G
Diethyl-aminomalonat Hydrochlorid 98%
Diethyl-aminomalonat Hydrochlorid
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₇H₁₄ClNO₄ Molecular Weight: 211,65 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012510 CAS-Nummer: 13433-00-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...