3-Amino-4-brom-1H-indazol 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD84125-250MGEA 27 EUR
BLDPBD84125-250MG BLDPBD84125-100MG BLDPBD84125-1G BLDPBD84125-5G
3-Amino-4-brom-1H-indazol 95%
3-Amino-4-bromo-1H-indazole

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₆BrN₃
Molecular Weight: 212,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09909580
CAS-Nummer: 914311-50-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR