3-Amino-3-(3-methoxyphenyl)propionsäure 95%

BLDPBD83939-1GEA 19 EUR
BLDPBD83939-1G BLDPBD83939-250MG
3-Amino-3-(3-methoxyphenyl)propionsäure 95%
3-Amino-3-(3-methoxyphenyl)propionsäure

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₀H₁₃NO₃
Molecular Weight: 195,22 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00462569
CAS-Nummer: 68208-19-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR