3-Amino-3-(2-fluorphenyl)propionsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD83876-1GEA 31 EUR
BLDPBD83876-1G BLDPBD83876-5G
3-Amino-3-(2-fluorphenyl)propionsäure 98%
3-Amino-3-(2-fluorphenyl)propionsäure

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₉H₁₀FNO₂
Molecular Weight: 183,1796 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00193241
CAS-Nummer: 117391-49-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR