3-Amino-2-chloro-6-methylphenol 98%

BLDPBD8343-100GEA 61 EUR
BLDPBD8343-100G BLDPBD8343-5G BLDPBD8343-25G
3-Amino-2-chloro-6-methylphenol 98%
3-Amino-2-chloro-6-methylphenol

Storage condition: Sealed in dry, 2-8°C

Formel: C₇H₈ClNO
Molecular Weight: 157,60 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03094650
CAS-Nummer: 84540-50-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR