4-(Trifluormethyl)salicylsäure 97%
Lieferant: BLD PHARMATECH GMBH
|
|
BLDPBD8328-10GEA
7
EUR
BLDPBD8328-10G
BLDPBD8328-1000G
BLDPBD8328-25G
BLDPBD8328-100G
BLDPBD8328-500G
BLDPBD8328-5G
4-(Trifluormethyl)salicylsäure 97%
4-(Trifluormethyl)salicylsäure
Storage condition: Sealed in dry, Room Temperature
Formel:
C₈H₅F₃O₃ Molecular Weight: 206,1187 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01529657 CAS-Nummer: 328-90-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...