1,3-Di-tert-butyltoluol 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD82989-100GEA 209 EUR
BLDPBD82989-100G BLDPBD82989-5G BLDPBD82989-10G BLDPBD82989-500G BLDPBD82989-1G BLDPBD82989-25G
1,3-Di-tert-butyltoluol 98%
1,3-Di-tert-butyltoluol

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₅H₂₄
Molecular Weight: 204,36 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00026300
CAS-Nummer: 15181-11-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR