4-Fluor-2-methylphenylboronsäure 98%
Lieferant: BLD PHARMATECH GMBH
|
|
BLDPBD8290-500GEA
651
EUR
BLDPBD8290-500G
BLDPBD8290-5G
BLDPBD8290-100G
BLDPBD8290-10G
BLDPBD8290-1G
BLDPBD8290-25G
4-Fluor-2-methylphenylboronsäure 98%
4-Fluor-2-methylphenylboronsäure
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel:
C₇H₈BFO₂ Molecular Weight: 153,95 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD17214439 CAS-Nummer: 139911-29-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...