2-Brom-3',5'-bis(trifluormethyl)acetophenon 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD82280-1GEA 41 EUR
BLDPBD82280-1G BLDPBD82280-250MG BLDPBD82280-5G
2-Brom-3',5'-bis(trifluormethyl)acetophenon 95%
2-Brom-3',5'-bis(trifluormethyl)acetophenon

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₀H₅BrF₆O
Molecular Weight: 335,0405 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00792434
CAS-Nummer: 131805-94-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR