3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydro-2-naphthalinboronsäure 95%

BLDPBD82224-5GEA 738 EUR
BLDPBD82224-5G BLDPBD82224-100MG BLDPBD82224-250MG BLDPBD82224-1G
3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydro-2-naphthalinboronsäure 95%
3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydro-2-naphthalinboronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₅H₂₃BO₂
Molecular Weight: 246,1529 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08436043
CAS-Nummer: 169126-64-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR