3,4-Diaminobenzhydrazid 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD81671-25GEA 359 EUR
BLDPBD81671-25G BLDPBD81671-0.025G BLDPBD81671-1G BLDPBD81671-5G
3,4-Diaminobenzhydrazid 98%
3,4-Diaminobenzhydrazid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₁₀N₄O
Molecular Weight: 166,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00017069
CAS-Nummer: 103956-09-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR