3,4,6-Trichlorpyridazin 97%

BLDPBD81616-25GEA 26 EUR
BLDPBD81616-25G BLDPBD81616-100G BLDPBD81616-10G BLDPBD81616-5G BLDPBD81616-500G
3,4,6-Trichlorpyridazin 97%
3,4,6-Trichloropyridazine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₄HCl₃N₂
Molecular Weight: 183,4231 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00834957
CAS-Nummer: 6082-66-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR