2-Formyl-5-methylphenylboronsäure 96%

BLDPBD8086-5GEA 867 EUR
BLDPBD8086-5G BLDPBD8086-1G BLDPBD8086-250MG
2-Formyl-5-methylphenylboronsäure 96%
2-Formyl-5-methylphenylboronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₉BO₃
Molecular Weight: 163,9663 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01319009
CAS-Nummer: 40138-17-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR