3-(4-Bromophenyl)prop-2-yn-1-ol 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD79954-5GEA 390 EUR
BLDPBD79954-5G BLDPBD79954-1G BLDPBD79954-100MG BLDPBD79954-250MG
3-(4-Bromophenyl)prop-2-yn-1-ol 97%
3-(4-Bromophenyl)prop-2-yn-1-ol

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₉H₇BrO
Molecular Weight: 211,055 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01250510
CAS-Nummer: 37614-58-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR