2-Amino-5-pyrimidinboronsäure 98%

BLDPBD7977-250MGEA 6 EUR
BLDPBD7977-250MG BLDPBD7977-5G BLDPBD7977-25G BLDPBD7977-10G BLDPBD7977-1G
2-Amino-5-pyrimidinboronsäure 98%
2-Amino-5-pyrimidineboronic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₄H₆BN₃O₂
Molecular Weight: 138,92 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07375147
CAS-Nummer: 936250-22-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR