3-(2,3-Dimethoxyphenyl)propionsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD79159-25GEA 164 EUR
BLDPBD79159-25G BLDPBD79159-250MG BLDPBD79159-1G BLDPBD79159-5G BLDPBD79159-0.025G
3-(2,3-Dimethoxyphenyl)propionsäure 98%
3-(2,3-Dimethoxyphenyl)propionsäure

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₁₄O₄
Molecular Weight: 210,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00029821
CAS-Nummer: 10538-48-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR