3-Amino-3-(2,3-dichlorphenyl)propionsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD79149-5GEA 213 EUR
BLDPBD79149-5G BLDPBD79149-1G BLDPBD79149-250MG
3-Amino-3-(2,3-dichlorphenyl)propionsäure 98%
3-Amino-3-(2,3-dichlorphenyl)propionsäure

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₉H₉Cl₂NO₂
Molecular Weight: 234,0800 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01871301
CAS-Nummer: 117391-56-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR