3-(2,4-Dimethoxyphenyl)propionsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD79129-1GEA 78 EUR
BLDPBD79129-1G BLDPBD79129-5G BLDPBD79129-250MG BLDPBD79129-10G
3-(2,4-Dimethoxyphenyl)propionsäure 98%
3-(2,4-Dimethoxyphenyl)propionsäure

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₁₄O₄
Molecular Weight: 210,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00598205
CAS-Nummer: 22174-29-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR