5-(1-Methylethyl)-2-pyridinamine 97%

BLDPBD78487-5GEA 597 EUR
BLDPBD78487-5G BLDPBD78487-1G BLDPBD78487-10G
5-(1-Methylethyl)-2-pyridinamine 97%
5-(1-Methylethyl)-2-pyridinamine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₈H₁₂N₂
Molecular Weight: 136,194 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08460246
CAS-Nummer: 603310-75-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR