2-Methylimidazo[1,2-a]pyridin-3-carbonsäure 95%

BLDPBD77276-250MGEA 24 EUR
BLDPBD77276-250MG BLDPBD77276-10G BLDPBD77276-5G BLDPBD77276-1G
2-Methylimidazo[1,2-a]pyridin-3-carbonsäure 95%
2-Methylimidazo[1,2-a]pyridin-3-carbonsäure

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₈N₂O₂
Molecular Weight: 176,1700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00244615
CAS-Nummer: 21801-79-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR