3'-Brom-4'-methylacetophenon 98%

BLDPBD7658-10GEA 53 EUR
BLDPBD7658-10G BLDPBD7658-1G BLDPBD7658-25G BLDPBD7658-5G BLDPBD7658-100G
3'-Brom-4'-methylacetophenon 98%
3'-Brom-4'-methylacetophenon

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₉BrO
Molecular Weight: 213,0700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02683854
CAS-Nummer: 40180-80-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR