Monomethylphthalat 95%
|
|
BLDPBD7642-100GEA
148
EUR
BLDPBD7642-100G
BLDPBD7642-5G
BLDPBD7642-25G
BLDPBD7642-10G
Monomethylphthalat 95%
Monomethylphthalat
Storage condition: Sealed in dry, Room Temperature
Formel:
C₉H₈O₄ Molecular Weight: 180,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002466 CAS-Nummer: 4376-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...