6-Benzyl-1-oxa-6-azaspiro[2.5]octane 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD763167-250MGEA 79 EUR
BLDPBD763167-250MG BLDPBD763167-1G BLDPBD763167-5G
6-Benzyl-1-oxa-6-azaspiro[2.5]octane 98%
6-Benzyl-1-oxa-6-azaspiro[2.5]octane

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₃H₁₇NO
Molecular Weight: 203,28 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06808592
CAS-Nummer: 19867-34-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR